Learn More
GE Healthcare Amersham™ ECL™ Western Blotting Detection Reagents: PROMO
Use for all routine Western blotting. GE Healthcare Life Sciences™ Amersham™ ECL™ Western Blotting Detection Reagents are the world’s first commercially available chemiluminescent detection reagents and still one of the most widely used.
Manufacturer:  GE Healthcare RPN2209PM
Catalog No. 45-000-885LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Detectable on Hyperfilm™ ECL or with CCD imager
  • Uses enhanced luminol-based detection which is suitable for all routine Western Blotting
Western Blotting
Specifications
Specifications
Routine Western blotting
125mL detection reagent, 1125mL detection reagent 2
≈10 pg
2°C to 8°C
125 mL
ECL western blotting detection reagents
Detection of 2000cm2 of membrane