Learn More
Cytiva Amersham™ Hybond™ -N+ Membranes
Use for the highest sensitivity in DNA and RNA blotting. Cytiva Life Sciences™ Amersham™ Hybond™ -N+ Membrane is a positively charged nylon membranes for use with radioactive and nonradioactive chemiluminescence and chemifluorescence detection.
$485.00 - $701.00
Specifications
For Use With (Application) Southern blots; UV fixation
Binding Capacity Up to 600μg/cm2
Material Nylon, Positively Charged
View More Specs
Products 8
Catalog Number Mfr. No. Dimensions (LxW) Quantity Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (LxW) Quantity Diameter (Metric) Price Quantity & Availability  
45-000-763
View Documents
Cytiva
RPN1210B
12 x 10 cm 20 Sheets Not Applicable
Each for $523.00
Add to Cart
45-000-850
View Documents
Cytiva
RPN203B
20 cm x 3 m 1 Roll Not Applicable
Each for $617.00
Add to Cart
45-000-927
View Documents
Cytiva
RPN303B
30 cm x 3 1 Roll Not Applicable
Each for $636.00
Add to Cart
45-000-949
View Documents
Cytiva
RPN82B
Not Applicable 50 Discs 82 mm
Each for $485.00
Add to Cart
45-000-761
View Documents
Cytiva
RPN119B
11.9 x 7.8 cm 50 Sheets Not Applicable
Each for $701.00
Add to Cart
45-000-785
View Documents
Cytiva
RPN1510B
15 x 10 cm 20 Sheets Not Applicable
Each for $523.00
Add to Cart
45-000-787
View Documents
Cytiva
RPN1520B
15 x 20 cm 10 Sheets Not Applicable
Each for $679.00
Add to Cart
45-000-838
View Documents
Cytiva
RPN2020B
20 x 20 cm 10 Sheets Not Applicable
Each for $599.00
Add to Cart
Description
Description
  • Pore size of 0.45μm with retention of nucleic acids >50bp
  • Binding capacity for nucleic acids 600μg/cm2
  • Suitable for high-density arrays
  • Excellent sensitivity in conventional Southern blotting
  • Nucleic acid samples may be fixed by UV or baking
  • Not suitable for alkali blotting and fixation
Specifications
Specifications
Southern blots; UV fixation
Up to 600μg/cm2
Nylon, Positively Charged