Learn More
GE Healthcare Amersham™ Hybond™ P 0.45µm PVDF Membrane PROMO
Use for chemiluminescent and fluorescent detection methods for proteins of > Mr 20 000. GE Healthcare Life Sciences™ Amersham™ Hybond™ P 0.45µm PVDF Membrane is a 0.45 µm pore size hydrophobic membrane, with high physical strength, highly suitable for stripping and reprobing.
Manufacturer:  GE Healthcare 29334865
Catalog No. 45-004-110PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Protein binding capacity over 200µg/cm2 for easy signal detection
  • High protein retention even after harsh washing steps
  • High protein capture during transfer minimizes sample loss
  • Available in popular precut sizes for your application
Specifications
Specifications
PVDF
0.45 μm
Western Blotting
>200 μg/cm2; IgG