Learn More
GE Healthcare Amersham™ Hybond™ PVDF Membranes: Rolls: PROMO
Ideal for Western blotting applications where stripping and reprobing are necessary.  GE Healthcare Life Sciences™ Amersham™ Hybond™ PVDF Membranes: Rolls  are hydrophobic PVDF membranes.
Products 1
Catalog Number Mfr. No. For Use With (Application) Grade Format Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Grade Format Price Quantity & Availability  
45-004-095LR View Documents GE Healthcare
10600022PM
Specifically optimized for fluorescence detection in Western blotting applications LFP 0.2μm Roll (260mm x 4m) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Supplied in rolls.
  • Hydrophobic PVDF membranes
  • Small pore size eliminates blow-through and increases protein binding over a wide range of molecular weights
Amersham Hybond 0.2μm
  • Compatible with a range of solvents used for rapid destaining
  • Amersham Hybond SEQ 0.2μm
    • Specifically designed for protein sequencing applications and solid phase amino acid analysis
    • Offers excellent blotting for proteins Mr<20,000
    Amersham Hybond LFP 0.2μm
    • Specifically optimized for fluorescence detection in Western blotting applications
    • Provides low background fluorescence, resulting in high sensitivity