Learn More
GE Healthcare Amersham™ Hyperfilm™ ECL: PROMO
Use with all blue and green light chemiluminescent systems. GE Healthcare Amersham™ Hyperfilm™ ECL is constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer.
Manufacturer:  GE Healthcare 28906835PM
Catalog No. 45-001-504LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  •  Antistatic coating reduces risk of visible artifacts
  • Clear base offers high contrast for easy reading of images
  • Suitable for use with all blue and green light chemiluminescent systems
  • Film can be processed in automatic processors or manually using most common X-ray film developers and fixers
  • Chemiluminescent detection in Western, Southern, and Northern blots
Specifications
Specifications
17.8 x 12.7 cm
50 Sheets/Box
Hyperfilm ECL
All blue and green light chemiluminescent system, chemiluminescent detection in western, southern, and northern blots
4°C