Learn More
GE Healthcare Amersham™ Hyperfilm™ MP: PROMO
Use in automatic processors or manually in tanks and trays using most common X-ray film developers and fixers. GE Healthcare Amersham™ Hyperfilm™ MP is an excellent multipurpose film constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti scratch layer.
Manufacturer:  GE Healthcare 28906842PM
Catalog No. 45-001-511LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • High sensitivity for low concentration targets
  • Optimized for direct and indirect autoradiography with 3H, 14C, 32P, 33P, 35S, 125I
  • Clear background for excellent contrast and improved band visibility.
  • Publication-quality images
  • High resolution at an economical price
  • Easy to use
Specifications
Specifications
17.8 x 12.7 cm
Detection of 3H, 14C, 32P, 33P, 35S, 125I in direct or indirect autoradiography, 32P, 33P, 125I southern, northern, and western blots; 32S, 33P, 350P sequencing, fluorography of 3H, 14C and 35S
50 Sheets/Box