Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ Megaprime™ DNA Labeling System

For labeling DNA probes to specific activities >1 × 109dpm/μg in 10 minutes

$1046.00 - $1533.00

Specifications

Content And Storage -15°C to -30°C
For Use With (Application) Membrane hybridization
View More Specs

Products 2
Catalog Number Mfr. No. Product Type No. of Reactions Includes Price Quantity & Availability  
Catalog Number Mfr. No. Product Type No. of Reactions Includes Price Quantity & Availability  
45-000-810
View Documents
Cytiva
RPN1607
Megaprime DNA Labeling System, dCTP (for use with labeled dCTP) 60 Primer solution 300μL, Labeling Buffered ATP, dGTP and dTTP in Tris HCl pH7.5, 2-mercaptoethanol and MgCl2 600μL, Nucleotide solutions dATP, cCTP, dGTP, dTTP in Tris HCl pH 8, 0.5mM EDTA, Reaction Buffer A 10 x concentr
Each for $1,533.00
Add to Cart
 
45-000-809
View Documents
Cytiva
RPN1606
Megaprime DNA Labeling System, dCTP (for use with labeled dCTP) 30 Primer solution 150μL, Labeling Buffer dATP, dGTP and dTTP in Tris HCl pH7.5, 2-mercaptoethanol and MgCl2 300μL, Nucleotide solutions dATP, cCTP, dGTP, dTTP in Tris HCl pH 8, 0.5mM EDTA, Reaction Buffer A 10 x concentra
Each for $1,046.00
Add to Cart
 
Description

Description

Megaprime™ DNA Labeling Systems are used for rapid, random-prime labeling from as little as 25ng substrate DNA.

  • Features cloned Klenow fragment and random nonamers in the random-prime labeling technique
  • No need to remove unincorporated nucleotides before hybridization
  • Protocols for labeling directly from low melting point agarose

Specifications

Specifications

-15°C to -30°C
Membrane hybridization
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit