Learn More
GE Healthcare Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls PROMO
Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. GE Healthcare Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Manufacturer:  GE Healthcare 29361665
Catalog No. 45-004-001PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Packed in rolls.
  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent
Specifications
Specifications
Roll
0.2 μm
30 cm
100% Pure Nitrocellulose
4 m
Western blots, dot-blot assays, and other protein or nucleic acid methods