Learn More
GE Healthcare Amersham™ Protran™ NC Nitrocellulose Western Blotting Sandwiches: PROMO
Manufacturer:  GE Healthcare 10600115PM
 View more versions of this product
Catalog No. 45-004-088LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Preassembled with two Whatman™ medium thickness 3MM Chr (0.34mm) filter papers to simplify set up
  • Fit most precase gel formats
Specifications
Specifications
Pack of 10
0.2 μm
9 cm
Sandwich
8 cm