Learn More
GE Healthcare Amersham™ Protran™ NC Nitrocellulose Western Blotting Sandwiches: PROMO
Manufacturer: GE Healthcare 10600116PM
Description
- Preassembled with two Whatman™ medium thickness 3MM Chr (0.34mm) filter papers to simplify set up
- Fit most precase gel formats
Specifications
Pack of 10 | |
0.1 μm | |
9 cm |
Sandwich | |
8 cm |