Learn More
GE Healthcare Amersham™ Protran™ NC Nitrocellulose Western Blotting Sandwiches: PROMO
Specifications
Format Sandwich, Pk. of 10
View More Specs
Products 3
Catalog Number Mfr. No. Length (Metric) Width (Metric) Pore Size Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Width (Metric) Pore Size Price Quantity & Availability  
45-004-087LR View Documents GE Healthcare
10600114PM
8cm 9cm 0.45μm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
45-004-088LR View Documents GE Healthcare
10600115PM
8cm 9cm 0.2μm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
45-004-089LR View Documents GE Healthcare
10600116PM
8cm 9cm 0.1μm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Preassembled with two Whatman™ medium thickness 3MM Chr (0.34mm) filter papers to simplify set up
  • Fit most precase gel formats
Specifications
Specifications
Sandwich, Pk. of 10