Learn More
GE Healthcare Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls PROMO
Use for the best results with fluorescence Western blotting detection applications. GE Healthcare Life Sciences™ Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Manufacturer:  GE Healthcare 29334867
Catalog No. 45-004-004PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Supplied in rolls.
  • Low background enables a dynamic range for complicated samples
  • Suitable for Western blots, dot-blot assays, and other protein or nucleic acid methods
  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
Specifications
Specifications
Roll
0.2 μm
30 cm
100% Pure Nitrocellulose
4 m
Western blots, dot-blot assays, and other protein or nucleic acid methods