Learn More
GE Healthcare Amersham™ Protran Supported
Manufacturer:  GE Healthcare 10600127
 View more versions of this product
Catalog No. 45-500-103
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Amersham Protran Supported offers excellent sensitivity, resolution, and low background, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western Blotting Systems. This robust support allows repeated stripping and reprobing. A robust protein blotting membrane, made of reinforced nitrocellulose for multiple reprobings.
  • High binding capacity
  • Versatile, robust, and excellent signal-to-noise ratios
  • Optimal for applications when stripping and reprobing is required
Specifications
Specifications
Blotting Membrane
Protein
1 Each
For western, southern and northern blotting applications