Learn More
GE Healthcare Amersham Protran Supported
Manufacturer: GE Healthcare 10600128
Description
Amersham Protran Supported offers excellent sensitivity, resolution, and low background, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western Blotting Systems. This robust support allows repeated stripping and reprobing. A robust protein blotting membrane, made of reinforced nitrocellulose for multiple reprobings.- High binding capacity
- Versatile, robust, and excellent signal-to-noise ratios
- Optimal for applications when stripping and reprobing is required
Specifications
Blotting Membrane | |
Protein | |
1 Each | |
For western, southern and northern blotting applications |