Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Amersham™ Protran Supported

Specifications

For Use With (Application) For western, southern and northern blotting applications
Quantity 1 Each
Sample Type Protein
Product Type Blotting Membrane
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
45-500-104
View Documents
GE Healthcare
10600128
Blotting Membrane This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
45-500-103
View Documents
GE Healthcare
10600127
Blotting Membrane This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Amersham Protran Supported offers excellent sensitivity, resolution, and low background, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western Blotting Systems. This robust support allows repeated stripping and reprobing. A robust protein blotting membrane, made of reinforced nitrocellulose for multiple reprobings.
  • High binding capacity
  • Versatile, robust, and excellent signal-to-noise ratios
  • Optimal for applications when stripping and reprobing is required
Specifications

Specifications

For western, southern and northern blotting applications
1 Each
Protein
Blotting Membrane
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit