Learn More
GE Healthcare Amersham™ Reinforced Nitrocellulose Blotting Membrane
Manufacturer:  GE Healthcare 10600126
 View more versions of this product
Catalog No. 45-500-102
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Low background and excellent signal-to-noise ratios.
  • Compatible with a variety of labeling and detection systems.
  • High binding capacity of small proteins (< Mr 10 000) reduces risk of ldquo;blow-throughrdquo;.
  • Proprietary nitrocellulose formula gives a long shelf life of bound proteins.
  • Available in popular precut sizes for your application.
Specifications
Specifications
Blotting Membrane
Protein
1 Each
For western, southern and northern blotting applications