Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva (Formerly GE Healthcare Life Sciences) Amersham™ Reinforced Nitrocellulose Blotting Membrane Available on GSA/VA Contract for Federal Government customers only.

$121.10

Specifications

For Use With (Application) For western, southern and northern blotting applications
Quantity 1 Each
Sample Type Protein
Product Type Blotting Membrane
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
45-500-100 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10600124
Blotting Membrane Pack of 10 for $121.10
Add to cart
 
45-500-102
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10600126
Blotting Membrane This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Amersham Protran offers excellent sensitivity, resolution, and low background, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western Blotting Systems. Nitrocellulose membrane.
  • Low background and excellent signal-to-noise ratios
  • Compatible with a variety of labeling and detection systems
  • High-binding capacity
  • Suitable for proteins of a wide range of molecular weights
  • Proprietary nitrocellulose formula gives a long shelf life of bound proteins
  • Available in popular precut sizes for your application

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

For western, southern and northern blotting applications
1 Each
Protein
Blotting Membrane
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit