Learn More
GE Healthcare Amersham™ Streptavidin-HRP Conjugate: PROMO
Product
Manufacturer:  GE Healthcare RPN1231100ULPM
Catalog No. 45-001-277LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • High specificity reagents
  • Conjugation achieved using the periodate coupling method described by Nakane and co-workers
  • Sold on the basis of titer in an ELISA and performance in a western blot
Specifications
Specifications
Suitable for use in ECL™, western blotting ,ELISA, immunocytochemistry
2mL of Phosphate Buffered Saline (0.1M Sodium Phosphate, 0.1M Sodium Chloride, pH 7.4) containing 1%(w/v) Bovine Serum Albumin and an antimicrobial agent
100mL