Learn More
EVOS™ EVOS™ Onstage Vessel Holder, One 35 mm Petri Dish
Supplier: EVOS™ AMEPVH029
Description
For use with EVOS FL Auto Digital Inverted Fluorescence System (Cat. No. 12-463-377).
Onstage Vessel Holders
- Accommodate a variety of flasks, plates and dishes
- Maximize your choice of cell culture vessels when conducting time-lapse imaging experiments using the EVOS FL Auto Imaging System equipped with onstage incubator
- Allow a perfect fit of your microwell plates or cell culture flasks/dishes to the master plate of the onstage incubator for increased precision in sample alignment
- With a snug and secure fit, yet readily switched, these vessel holders make time-lapse imaging convenient and easy
Specifications
Vessel Holder | |
EVOS™ FL Auto Imaging System | |
1/Ea. |
EVOS | |
Holds one 35 mm Petri dish |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.