Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ AMG PERK 44
Potent and selective PERK inhibitor; orally bioavailable
$402.00 - $1622.00
Specifications
Chemical Name or Material | 4-[2-Amino-4-methyl-3-(2-methyl-6-quinolinyl)benzoyl]-1,2-dihydro-1-methyl-2,5-diphenyl-3H-pyrazol-3-one hydrochloride |
---|---|
CAS | 1883548-84-2 |
Target | PERK Inhibitors |
Molecular Formula | C34H28N4O2.HCl |
Formula Weight | 561.07 |
Description
Potent and selective PERK inhibitor (IC50 = 6 nM). Exhibits >160-fold selectivity for PERK over B-Raf and GCN2, as well as a panel of 387 other kinases. Orally bioavailable.Specifications
4-[2-Amino-4-methyl-3-(2-methyl-6-quinolinyl)benzoyl]-1,2-dihydro-1-methyl-2,5-diphenyl-3H-pyrazol-3-one hydrochloride | |
PERK Inhibitors | |
561.07 |
1883548-84-2 | |
C34H28N4O2.HCl | |
>98% |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title