Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ AMICA/JAML Antibody Pair [HRP]
Mouse Monoclonal Antibody Pair
$1183.00
Specifications
Assay Range | 234.375 pg/mL |
---|---|
Assay Sensitivity | 234.38-15000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB009925
|
Novus Biologicals™
NBP27944315PLATES |
15 Plates |
Each of 1 for $1,183.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human AMICA/JAML.Specifications
234.375 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638 | |
JAML | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 15 ng/mL is recommended. |
234.38-15000 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human AMICA/JAML. | |
Sandwich ELISA | |
120425 | |
Mouse | |
RUO | |
AMICA/JAML |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title