Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMICA/JAML Armenian Hamster anti-Mouse, DyLight™ 550, Clone: 40000000000, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143309R

Catalog No. NBP143309R


Add to cart

Description

Description

AMICA/JAML Monoclonal antibody specifically detects AMICA/JAML in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

AMICA/JAML
40000000000
Flow Cytometry
Conjugated, Purified
JAML
Synthetic peptide corresponding to residues C(267) D G I L G D N F R P T Q(279) of mouse CARP.
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Mouse
Flow Cytometry
DyLight™ 550
50 mM sodium borate with 0.05% sodium azide
adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638
Armenian Hamster
IgG
0.1 mL
Primary
120425.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit