Learn More
MilliporeSigma™ Amicon™ Pro Purification System with Ultra-0.5 Device, 10 Kda
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
Manufacturer:  MilliporeSigma™ ACS501002
 View more versions of this product
Catalog No. ACS501002
Add to cart
Description
Description
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
The Amicon™ Pro Purification System includes: Amicon Ultra NMWCO filters, Exchange devices, and Collection tubes with cap.
Specifications
Specifications
15 mL
10 KDa
Regenerated Cellulose (RC)
12 cm
2Pack
Yes
Non-sterile
Centrifugal Filters
2 Amicon Ultra NMWCO 10 kDa filters, 2 Exchange devices, and 2 Collection tubes with cap