Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Amicon™ Pro Purification System with Ultra-0.5 Device, 30 Kda
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
Supplier: MilliporeSigma™ ACS503002
Description
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
The Amicon™ Pro Purification System includes: Amicon Ultra NMWCO filters, Exchange devices, and Collection tubes with cap.Specifications
Single-use | |
Non-sterile | |
2 Amicon Ultra NMWCO 30 KDa filters, 2 Exchange devices, and 2 Collection tubes with cap | |
Centrifugal Filters | |
12 cm |
30 kDa | |
Regenerated Cellulose (RC) | |
2Pack | |
10 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction