Learn More
MilliporeSigma™ Amicon™ Pro Purification System with Ultra-0.5 Device, 30 Kda
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
$30.35 - $304.50
Products 3
Catalog Number Mfr. No. Includes Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Includes Quantity Price Quantity & Availability  
ACS503024
View Documents
MilliporeSigma™
ACS503024
24 Amicon Ultra NMWCO 30 KDa filters, 24 Exchange devices, and 24 Collection tubes with cap 24Pack
Pack of 24 for $304.50
Add to cart
ACS503012
View Documents
MilliporeSigma™
ACS503012
12 Amicon Ultra NMWCO 30 KDa filters, 12 Exchange devices, and 12 Collection tubes with cap 12Pack
Pack of 12 for $195.60
Add to cart
ACS503002
View Documents
MilliporeSigma™
ACS503002
2 Amicon Ultra NMWCO 30 KDa filters, 2 Exchange devices, and 2 Collection tubes with cap 2Pack
Pack of 2 for $30.35
Add to cart
Description
Description
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
The Amicon™ Pro Purification System includes: Amicon Ultra NMWCO filters, Exchange devices, and Collection tubes with cap.