Learn More
MilliporeSigma™ Amicon™ Pro Purification System with Ultra-0.5 Device, 50 Kda
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
Supplier: MilliporeSigma™ ACS505012
Description
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
The Amicon™ Pro Purification System includes: Amicon Ultra NMWCO filters, Exchange devices, and Collection tubes with cap.Specifications
Single-use | |
Non-sterile | |
12 Amicon Ultra NMWCO 50 KDa filters, 12 Exchange devices, and 12 Collection tubes with cap | |
Centrifugal Filters |
50 kDa | |
Regenerated Cellulose (RC) | |
12Pack | |
12 cm |