Learn More
Abnova™ AMID 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00084883T02
Catalog No. 89-933-304
Add to cart
Description
Description
Specifications
Specifications
293T
NM_032797
pCMV-AMID full-length
100 μL
Human
AMID 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 40.5kDa
Transient overexpression cell lysate was tested with anti-AMID antibody (Mfr. No. H00084883-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot