Alfa Aesar™ Amidosulfonic acid, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4451022

Quantity 100g

 View more versions of this product

Catalog No. AA4451022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5329-14-6
    Molecular Formula H2NSO3H
    Formula Weight 97.09
    MDL Number MFCD00011603
    UN Number UN2967
    Merck Index 14,8921
    Chemical Name or Material Amidosulfonic acid

    Specifications

    Formula Weight 97.09
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    Molecular Formula H2NSO3H
    CAS 5329-14-6
    Melting Point ≈205°C (dec.)
    RTECS Number W05950000
    Odor Odorless
    TSCA TSCA
    pH 1.2
    Density 2.12g/mL
    MDL Number MFCD00011603
    Merck Index 14,8921
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: SULFAMIC ACID
    Physical Form Crystalline
    UN Number UN2967
    Quantity 100g
    Chemical Name or Material Amidosulfonic acid
    Assay 99.99%, (metals basis)

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit