Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMIGO3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 212781AP150UL
Description
AMIGO3 may mediate heterophilic cell-cell interaction. AMIGO3 may contribute to signal transduction through its intracellular domain.Specifications
AMIGO3 | |
Polyclonal | |
Unconjugated | |
AMIGO3 | |
ALI3, Alivin 3, AMIGO 3, AMIGO3, Amphoterin induced protein 3, KIAA1851 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
316003, 320844, 386724 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q80ZD5, Q86WK7, Q8C2S7 | |
Amigo3 | |
AMIGO3 Fusion Protein Ag15790 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction