Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amikacin (From Kanamycin A, White Crystalline Powder), Fisher BioReagents

Catalog Number bp2643250
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass
This item is not returnable. View return policy
37517-28-5
C22H43N5O13
585.608
LKCWBDHBTVXHDL-RMDFUYIESA-N
Lukadin
37768
CHEBI:2637
(2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy-3-hydroxycyclohexyl]-2-hydroxybutanamide
C1C(C(C(C(C1NC(=O)C(CCN)O)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(C(C(C(O3)CN)O)O)O)N
This item is not returnable. View return policy
37517-28-5
C22H43N5O13
585.608
LKCWBDHBTVXHDL-RMDFUYIESA-N
Lukadin
37768
CHEBI:2637
(2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy-3-hydroxycyclohexyl]-2-hydroxybutanamide
C1C(C(C(C(C1NC(=O)C(CCN)O)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(C(C(C(O3)CN)O)O)O)N
  • An antibiotic that serves as a selective agent to isolate transformed bacteria
  • Amikacin functions as an indirect inhibitor of protein synthesis in prokaryotes by binding to the 30S subunit of ribosomes
  • Useful as a selective agent to isolate bacteria that have been transformed with plasmid containing the antibiotic-resistant gene
  • Intended for research purposes only; not for manufacturing or commercial use
TRUSTED_SUSTAINABILITY
Type Amikacin
Physical Form Solid
pH 9.5 to 11.5
Melting Point 214°C
Merck Index 15, 400
Packaging Amber Glass
Potency > 900μg/mg
Solubility Information completely soluble in water
Formula Weight 585.61
Color White
Quantity 250 mg
Chemical Name or Material Amikacin, From Kanamycin A
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
TSCA Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.
Recommended Storage 0° to 5°C, desiccate.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit