Learn More
Amino Acids Small Model Set Products for Science Education
Model of each of the common 20 alpha amino acids.
Manufacturer:  Ryler Enterprises AA1
Catalog No. S94025
Add to cart
Includes: Kit Contents:
  • 20 white 1-peg hydrogen atom centers
  • 4 blue 4-peg nitrogen atom centers
  • 7 red 2-peg oxygen atom centers
  • 2 yellow 2-peg sulfur atom centers
  • 9 black 4-peg carbon atom centers
  • 32-1.25 in. PVC, clear bonds (single bonds)
  • 7-1.625 in. PVC, clear bonds (double bonds)
  • 3-2 in. PVC, white bonds (H bonds)
  • Lab procedure
  • Answer key
Description
Description
Students can build three amino acids simultaneously with this kit to investigate D and L isomers of amino acids, hydrogen bonding, dehydration synthesis, peptide formation, the meaning of “alpha amino acid”, primary and secondary structures of proteins, and amines such as GABA and epinephrine. As separate investigations, students can make models of each of the common 20 alpha amino acids.
  • Color-coded, modeled plastic atom centers are 1/4 in. diameter
  • Pegs are 3/8 in. long and are set at angles peculiar to the elements the atoms represent
  • Standard color-coding
  • Atom centers are bonded by connecting pegs with pvc tubing
Specifications
Specifications
To investigate D and L forms of Amino Acids, Hydrogen bonding, dehydration synthesis
20 White 1-peg Hydrogen atom centers, 4 blue 4-peg Nitrogen atom centers, 7 red 2-peg Oxygen atom centers, 2 yellow 2-peg sulfur atom centers, 9 black 4-peg Carbon atom centers, Thirty two 1.25 in. PVC, clear bonds (single bonds), Seven 1.625 in. PVC, clear bonds (double bonds), 3 to 2 in. PVC, white bonds (H bonds), lab procedure, answer key
Amino Acid small model set