Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminoacetonitrile Hydrogen Sulfate 97.0+%, TCI America™

Click to view available options
25G
151-63-3
C2H6N2O4S
154.14
MFCD00012851
GTGIXCPOLMWQTC-UHFFFAOYSA-N
aminoacetonitrile bisulfate,2-aminoacetonitrile sulfate,aminoacetonitrile hydrogen sulfate,aminoacetonitrile sulfate 1:1,aminoacetonitrile hydrogensulfate,2-aminoacetonitrile; sulfuric acid,aminoacetonitrile bisulphate,unii-7xa82hgq1c,aminoacetonitrile hydrosulfate,aminomethylcyanide hydrogen sulfate
196971
2-aminoacetonitrile;sulfuric acid
C(C#N)N.OS(=O)(=O)O
151-63-3
C2H6N2O4S
154.14
MFCD00012851
GTGIXCPOLMWQTC-UHFFFAOYSA-N
aminoacetonitrile bisulfate,2-aminoacetonitrile sulfate,aminoacetonitrile hydrogen sulfate,aminoacetonitrile sulfate 1:1,aminoacetonitrile hydrogensulfate,2-aminoacetonitrile; sulfuric acid,aminoacetonitrile bisulphate,unii-7xa82hgq1c,aminoacetonitrile hydrosulfate,aminomethylcyanide hydrogen sulfate
196971
2-aminoacetonitrile;sulfuric acid
C(C#N)N.OS(=O)(=O)O
Melting Point 120°C
Quantity 25g
Color White-Yellow
UN Number 3439
Formula Weight 154.14
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (N,T)
Chemical Name or Material Aminoacetonitrile Hydrogen Sulfate

EINECSNumber : (2)-1519

RTECSNumber : AL7800000

TSCA : No