Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Natamycin, Honeywell Fluka™

Click to view available options
Quantity:
50 mg
7681-93-8
7681-93-8

Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
Grade Analytical Standard
Packaging Glass Bottle
Quantity 50 mg
For Use With (Application) ANTIBIOTICS