Alfa Aesar™ (Aminoethylaminomethyl)phenethyltrimethoxysilane, mixture of m and p isomers

As a crosslinking agent

Manufacturer: Alfa Aesar™ 4390404

Quantity 2g

 View more versions of this product

Catalog No. AA4390404

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 74113-77-2
    Molecular Formula H2NC2H4NHCH2C6H4C2H4Si(OCH3)3
    Formula Weight 298.46
    MDL Number MFCD00054775
    UN Number UN2735
    Chemical Name or Material (Aminoethylaminomethyl)phenethyltrimethoxysilane, Mixture of m and p isomers

    Specifications

    Formula Weight 298.46
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula H2NC2H4NHCH2C6H4C2H4Si(OCH3)3
    Boiling Point 128°C (0.2mmHg)
    Color Colorless to pale yellow
    Quantity 2g
    Physical Form Liquid
    UN Number UN2735
    Flash Point >110°C (230°F)
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Sensitivity Air & Moisture Sensitive
    MDL Number MFCD00054775
    CAS 74113-77-2
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Density 1.02g/mL
    Chemical Name or Material (Aminoethylaminomethyl)phenethyltrimethoxysilane, Mixture of m and p isomers

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit