Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminoglutethimide, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Amber Glass Bottle
125-84-8
C13H16N2O2
232.28
ROBVIMPUHSLWNV-UHFFFAOYNA-N
3-(4-aminophenyl)-3-ethylpiperidine-2,6-dione
CCC1(CCC(=O)NC1=O)C1=CC=C(N)C=C1
125-84-8
C13H16N2O2
232.28
ROBVIMPUHSLWNV-UHFFFAOYNA-N
3-(4-aminophenyl)-3-ethylpiperidine-2,6-dione
CCC1(CCC(=O)NC1=O)C1=CC=C(N)C=C1

A1139, 125-84-8, C13H16N2O2

Spectrum™ Aminoglutethimide, USP is an anti-steroid drug and is clinically used in the treatment of Cushing's syndrome. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.001
Loss on Drying 0.005
Quantity 5 kg
pH 6.2 to 7.3
CAS Max % 1
Percent Purity 98 to 102%