Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aminoguanidine bicarbonate, 98.5%, ACROS Organics™
Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic Bottle
Plastic drum
Specifications
Infrared Spectrum | Authentic |
Melting Point | 125°C |
Residue after Ignition | 0.3% max |
Color | White |
Quantity | 250g |
Packaging | Plastic Bottle |
Ignition Residue | 0.3% max. |
Assay Percent Range | 98.5% |
Linear Formula | H2NNHC(=NH)NH2·H2CO3 |
Beilstein | 03,117 |
Show More |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
May damage fertility or the unborn child.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention
GHS Signal Word: Danger
EINECSNumber : 219-956-7