Alfa Aesar™ Aminoguanidine hydrogen carbonate, 98+%

Manufacturer: Alfa Aesar™ B2469622

Quantity 100g

 View more versions of this product

Catalog No. AAB2469622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2582-30-1
    Molecular Formula CH6N4·H2CO3
    Formula Weight 136.11
    MDL Number MFCD00012949
    UN Number UN3077
    Beilstein 3569869
    Chemical Name or Material Aminoguanidine hydrogen carbonate

    Specifications

    Formula Weight 136.11
    TSCA TSCA
    Quantity 100g
    RTECS Number FG1772000
    Molecular Formula CH6N4·H2CO3
    Melting Point ≈170°C (dec.)
    MDL Number MFCD00012949
    CAS 2582-30-1
    UN Number UN3077
    Beilstein 3569869
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H317
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material Aminoguanidine hydrogen carbonate
    Assay ≥98%

    Description

    Applications
    Aminoguanidine bicarbonate is an inhibitor of NOS (nitric oxide synthase). Aminoguanidine bicarbonate is used to study the effect of addition of polyamines to rat embryo cell cultures infected with adenovirus type 5.

    Solubility
    Soluble in water (3.3 mg/ml at 30°C), and water (2.7 mg/ml at 20°C).

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H317
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit