Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aminoguanidine hydrogen carbonate, 98+%

Click to view available options
Quantity:
500g
2500g
2582-30-1
C2H8N4O3
136.111
MFCD00012949
OTXHZHQQWQTQMW-UHFFFAOYSA-N
aminoguanidine bicarbonate,aminoguanidine hydrogen carbonate,aminoguanidinium bicarbonate,aminoguanidine hydrocarbonate,aminoguanidine carbonate 1:1,aminoguanidium hydrogen carbonate,n1-aminoguanidine carbonate 1:1,aminoguanidine hydrogencarbonate,aminoguanidinebicarbonate,hydrazinecarboximidamide carbonate
164944
2-aminoguanidine;carbonic acid
C(=NN)(N)N.C(=O)(O)O
2582-30-1
C2H8N4O3
136.111
MFCD00012949
OTXHZHQQWQTQMW-UHFFFAOYSA-N
aminoguanidine bicarbonate,aminoguanidine hydrogen carbonate,aminoguanidinium bicarbonate,aminoguanidine hydrocarbonate,aminoguanidine carbonate 1:1,aminoguanidium hydrogen carbonate,n1-aminoguanidine carbonate 1:1,aminoguanidine hydrogencarbonate,aminoguanidinebicarbonate,hydrazinecarboximidamide carbonate
164944
2-aminoguanidine;carbonic acid
C(=NN)(N)N.C(=O)(O)O
Applications
Aminoguanidine bicarbonate is an inhibitor of NOS (nitric oxide synthase). Aminoguanidine bicarbonate is used to study the effect of addition of polyamines to rat embryo cell cultures infected with adenovirus type 5.

Solubility
Soluble in water (3.3 mg/ml at 30°C), and water (2.7 mg/ml at 20°C).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Melting Point ~170°C (decomposition)
Quantity 500g
UN Number UN3077
Beilstein 3569869
Formula Weight 136.11
Percent Purity ≥98%
Chemical Name or Material Aminoguanidine hydrogen carbonate

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : FG1772000

TSCA : TSCA