Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aminoguanidine sulfate, 98%

Click to view available options
Quantity:
1000g
1068-42-4
CH8N4O4S
172.159
MFCD00151369
XLONNWGCEFSFTN-UHFFFAOYSA-N
aminoguanidine sulfate,guanylhydrazinesulfate,aminoguanidine sulphate,carbazamidine sulfate,2-aminoguanidine; sulfuric acid,amino-guanidine; sulfuric acid,aminoguanidinium sulfate,1-aminoguanidinium sulfate,ch6n4.h2so4,spectrum1505162
114832
2-aminoguanidine;sulfuric acid
C(=NN)(N)N.OS(=O)(=O)O
1068-42-4
CH8N4O4S
172.159
MFCD00151369
XLONNWGCEFSFTN-UHFFFAOYSA-N
aminoguanidine sulfate,guanylhydrazinesulfate,aminoguanidine sulphate,carbazamidine sulfate,2-aminoguanidine; sulfuric acid,amino-guanidine; sulfuric acid,aminoguanidinium sulfate,1-aminoguanidinium sulfate,ch6n4.h2so4,spectrum1505162
114832
2-aminoguanidine;sulfuric acid
C(=NN)(N)N.OS(=O)(=O)O
Quantity 1000g
Formula Weight 246.24
Percent Purity 98%
Chemical Name or Material Aminoguanidine sulfate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 213-945-0

TSCA : No

Recommended Storage : Ambient temperatures