Alfa Aesar™ Aminomethanesulfonic acid, 97%

Manufacturer: Alfa Aesar™ B2344136

Quantity 500g

 View more versions of this product

Catalog No. AAB2344136

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13881-91-9
    Molecular Formula H2NCH2SO3H
    Formula Weight 111.12
    MDL Number MFCD00008124
    UN Number UN2585
    Beilstein 1811756
    Synonym Aminomethylsulfonic acid
    Chemical Name or Material Aminomethanesulfonic acid

    Specifications

    Formula Weight 111.12
    TSCA TSCA
    Melting Point ≈180°C (dec.)
    Quantity 500g
    MDL Number MFCD00008124
    UN Number UN2585
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    pH 3.2
    Molecular Formula H2NCH2SO3H
    CAS 13881-91-9
    Beilstein 1811756
    Synonym Aminomethylsulfonic acid
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
    Chemical Name or Material Aminomethanesulfonic acid
    Assay 97%

    Description

    Applications
    Aminomethanesulfonic acid is used as a pharmaceutical intermediates.

    Solubility
    Soluble in water. (17 g/L) at 17°C

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit