Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminomethyl Polystyrene Resin cross-linked with 1% DVB (200-400mesh) (2.0-3.0mmol/g), TCI America™

Click to view available options
Quantity:
5g
25g
MFCD00130502
MFCD00130502
Color Yellow
Quantity 25g
Physical Form Crystal-Powder at 20°C
Chemical Name or Material Aminomethyl Polystyrene Resin cross-linked with 1% DVB (200-400mesh) (2.0-3.0mmol/g)