Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminomethylepolymer resin, 1% crossl. with DVB, 0.9-1.2mmol/g, 200-400 mesh, Acros Organics

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
78578-28-6
C19H21N
263.384
MFCD01092588
NBQVIZAFOXHTAI-UHFFFAOYSA-N
131636715
1,2-bis(ethenyl)benzene;(4-ethenylphenyl)methanamine
C=CC1=CC=C(C=C1)CN.C=CC1=CC=CC=C1C=C
78578-28-6
C19H21N
263.384
MFCD01092588
NBQVIZAFOXHTAI-UHFFFAOYSA-N
131636715
1,2-bis(ethenyl)benzene;(4-ethenylphenyl)methanamine
C=CC1=CC=C(C=C1)CN.C=CC1=CC=CC=C1C=C
for solid support synthesis
Additional Information crosslinking: 1% DVB
Loading 0.9 to 1.2mmol/g
Mesh Size 200 to 400 mesh
Packaging Glass bottle
Quantity 25g
Color White to Brown
Physical Form Beads
Chemical Name or Material Aminomethylepolymer resin, 1% crossl. with DVB, 0.9-1.2 mmol/g

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.