Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminomethyltransferase Antibody (OTI6F4), Alexa Fluor™ 405, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270171AF405

Catalog No. N270171A405


Only null left
Add to Cart

Description

Description

Aminomethyltransferase Monoclonal specifically detects Aminomethyltransferase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Aminomethyltransferase
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
AMT
Recombinant protein fragment corresponding to amino acids 146-403 of human Aminomethyltransferase (NP_000472) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Immunohistochemistry
OTI6F4
Western Blot, Immunohistochemistry
aminomethyltransferase, EC 2.1.2.10, GCE, GCSTaminomethyltransferase (glycine cleavage system protein T), glycine cleavage system protein T, NKHmitochondrial
Mouse
Affinity Purified
Neuroscience
275.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.