Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminophylline, Anhydrous, Powder, BP, EP, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
317-34-0
C16H24N10O4
420.43
FQPFAHBPWDRTLU-UHFFFAOYSA-N
bis(1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione); ethane-1,2-diamine
NCCN.CN1C2=C(NC=N2)C(=O)N(C)C1=O.CN1C2=C(NC=N2)C(=O)N(C)C1=O
317-34-0
C16H24N10O4
420.43
FQPFAHBPWDRTLU-UHFFFAOYSA-N
bis(1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione); ethane-1,2-diamine
NCCN.CN1C2=C(NC=N2)C(=O)N(C)C1=O.CN1C2=C(NC=N2)C(=O)N(C)C1=O

A4086, 317-34-0, C16H24N10O4

Spectrum™ Aminophylline, Anhydrous, Powder, BP, EP
Packaging Amber Glass Bottle
Water 0.015
Sulfated Ash 0.001
Quantity 100 g
CAS Max % 1
UN Number UN2811