Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminosalicylic Acid, USP, 98.5-100.5%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
50 kg
Packaging:
Amber Glass Bottle
65-49-6
C7H7NO3
153.14
WUBBRNOQWQTFEX-UHFFFAOYSA-N
4-amino-2-hydroxybenzoic acid
NC1=CC=C(C(O)=O)C(O)=C1
65-49-6
C7H7NO3
153.14
WUBBRNOQWQTFEX-UHFFFAOYSA-N
4-amino-2-hydroxybenzoic acid
NC1=CC=C(C(O)=O)C(O)=C1

AM167, 65-49-6, C7H7NO3

Spectrum™ Aminosalicylic Acid, USP , also known as 4-Aminosalicylic acid or para-aminosalicylic acid (PAS), is an antibiotic primarily used to treat active drug resistant tuberculosis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.2%
Water 0.5%
Packaging Amber Glass Bottle
Quantity 100 g
CAS Max % 100%
Percent Purity 98.5 to 100.5%