Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amiodarone hydrochloride, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
19774-82-4
C25H29I2NO3·HCl
681.77
ITPDYQOUSLNIHG-UHFFFAOYSA-N
amiodarone hydrochloride,amiodarone hcl,amiodar,nexterone,pacerone,amiodaronum hydrochloride,ritmocardyl,rythmarone,angoron
441325
(2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride
CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl
19774-82-4
C25H29I2NO3·HCl
681.77
ITPDYQOUSLNIHG-UHFFFAOYSA-N
amiodarone hydrochloride,amiodarone hcl,amiodar,nexterone,pacerone,amiodaronum hydrochloride,ritmocardyl,rythmarone,angoron
441325
(2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride
CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl
Melting Point 156.0°C
Quantity 25g
Infrared Spectrum Authentic
Loss on Drying 0.5% max.
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: insoluble.
Formula Weight 681.77
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Amiodarone hydrochloride

GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging the unborn child.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position

GHS Signal Word: Warning

EINECSNumber : 243-293-2