Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amiodarone Hydrochloride, EP, 98.5-101%, Spectrum™

Click to view available options
Quantity:
5 kg
25 kg
Packaging:
Fiber Drum
Poly Pail
19774-82-4
C25H30ClI2NO3
681.78
ITPDYQOUSLNIHG-UHFFFAOYSA-N
hydrogen {2-[4-(2-butyl-1-benzofuran-3-carbonyl)-2,6-diiodophenoxy]ethyl}diethylamine chloride
[H+].[Cl-].CCCCC1=C(C(=O)C2=CC(I)=C(OCCN(CC)CC)C(I)=C2)C2=CC=CC=C2O1
19774-82-4
C25H30ClI2NO3
681.78
ITPDYQOUSLNIHG-UHFFFAOYSA-N
hydrogen {2-[4-(2-butyl-1-benzofuran-3-carbonyl)-2,6-diiodophenoxy]ethyl}diethylamine chloride
[H+].[Cl-].CCCCC1=C(C(=O)C2=CC(I)=C(OCCN(CC)CC)C(I)=C2)C2=CC=CC=C2O1

A1189, 19774-82-4, C25H29I2NO3•HCl

Spectrum™ Amiodarone Hydrochloride, EP is an antiarrhythmic medication used to treat and prevent a number of types of irregular heartbeats. It is on the World Health Organization′s List of Essential Medicines. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Pail
Loss on Drying 0.005
Sulfated Ash 0.001
Quantity 5 kg
CAS Max % 1
Percent Purity 98.5 to 101%