Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amitriptyline Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
10 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
549-18-8
C20H24ClN
313.87
KFYRPLNVJVHZGT-UHFFFAOYSA-N
hydrogen dimethyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene}propyl)amine chloride
[H+].[Cl-].CN(C)CCC=C1C2=CC=CC=C2CCC2=CC=CC=C12
549-18-8
C20H24ClN
313.87
KFYRPLNVJVHZGT-UHFFFAOYSA-N
hydrogen dimethyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene}propyl)amine chloride
[H+].[Cl-].CN(C)CCC=C1C2=CC=CC=C2CCC2=CC=CC=C12

A1322, 549-18-8, C20H23N•HCl

Spectrum™ Amitriptyline Hydrochloride, USP is a medication of the tricyclic antidepressant class that is used for the treatment of mental illnesses. It is on the World Health Organization′s list of essential medicines. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.001
Loss on Drying 0.005
Quantity 5 g
CAS Max % 1
UN Number UN2811
Percent Purity 98 to 102%