Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amlodipine Besylate, EP, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
10 kg
Packaging:
Amber Glass Bottle
Fiber Drum
111470-99-6
C26H31ClN2O8S
567.05
ZPBWCRDSRKPIDG-UHFFFAOYNA-N
3-ethyl 5-methyl 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate; benzenesulfonic acid
OS(=O)(=O)C1=CC=CC=C1.CCOC(=O)C1=C(COCCN)NC(C)=C(C1C1=CC=CC=C1Cl)C(=O)OC
111470-99-6
C26H31ClN2O8S
567.05
ZPBWCRDSRKPIDG-UHFFFAOYNA-N
3-ethyl 5-methyl 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate; benzenesulfonic acid
OS(=O)(=O)C1=CC=CC=C1.CCOC(=O)C1=C(COCCN)NC(C)=C(C1C1=CC=CC=C1Cl)C(=O)OC

A2196, 111470-99-6, C20H25ClN2O5•C6H6O3S

Spectrum™ Amlodipine Besylate, EP is a medication used to treat high blood pressure and coronary artery disease. It is on the World Health Organization's List of Essential Medicines. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Appearance White to Off-White Crystalline powder
Quantity 25 g
CAS Max % 1