Learn More
Abnova™ AMMECR1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00009949T01
Catalog No. 89-933-313
Add to cart
Description
Description
Specifications
Specifications
293T
NM_001025580.1
pCMV-AMMECR1 full-length
100 μL
Human
AMMECR1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 32.67kDa
Transient overexpression cell lysate was tested with anti-AMMECR1 antibody (Mfr. No. H00009949-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot